Phosphoenolpyruvic acid monocyclohexylammonium salt ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P0758-1G P0758-100MG
TCIAP0758-1GEA 75.9 EUR
TCIAP0758-1G TCIAP0758-100MG
Phosphoenolpyruvic acid monocyclohexylammonium salt ≥98.0% (by titrimetric analysis)
Phosphoenolpyruvic acid monocyclohexylammonium salt
Formule: C₃H₅O₆P
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00036375
CAS nummer: 10526-80-4
EINECS: 234-084-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR