(R)-(+)-α-Methylbenzylamine ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

P0794-25ML P0794-500ML P0794-100ML
TCIAP0794-25MLEA 33 EUR
TCIAP0794-25ML TCIAP0794-500ML TCIAP0794-100ML
(R)-(+)-α-Methylbenzylamine ≥99.0% (by GC)
(R)-(+)-α-Methylbenzylamine
Formule: C₈H₁₁N
Kookpunt: 85°C
Dichtheid: 0,95 g/cm³ (20 °C)
Vlampunt: 70°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064405
CAS nummer: 3886-69-9
EINECS: 223-423-4
UN: 2922
ADR: 8,II
Merck Index: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9074e348f042639b9094ac142701e2f83761b174&vid=9074e348f042639b9094ac142701e2f83761b174&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %
Specific rotation [a]20/D +37.0 to +41.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR