(R)-(+)-1-Phenylethanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

P0795-1G P0795-25G P0795-5G
TCIAP0795-1GEA 45.1 EUR
TCIAP0795-1G TCIAP0795-25G TCIAP0795-5G
(R)-(+)-1-Phenylethanol ≥98.0% (by GC)
(R)-(+)-1-Phenylethanol
Formule: C₈H₁₀O
Kookpunt: 207 °C (1013 hPa)
Dichtheid: 1,012 g/cm³ (20 °C)
Vlampunt: 86°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064263
CAS nummer: 1517-69-7
UN: 2937
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1193df44239755dc9a822136ae1701f3307f5aa&vid=e1193df44239755dc9a822136ae1701f3307f5aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR