(±)-2-Phenyl-1-propanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

P0813-25ML P0813-500ML
TCIAP0813-25MLEA 25.3 EUR
TCIAP0813-25ML TCIAP0813-500ML
(±)-2-Phenyl-1-propanol ≥98.0% (by GC)
(±)-2-Phenyl-1-propanol
Formule: C₆H₅CH(CH₃)CH₂OH
Kookpunt: 223°C
Smeltpunt: –37 °C
Dichtheid: 1 g/cm³ (20 °C)
Vlampunt: 94°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004736
CAS nummer: 1123-85-9
EINECS: 214-379-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ed8a69c92257353cb64c06069f202e9139344b5&vid=0ed8a69c92257353cb64c06069f202e9139344b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR