N,N,N',N',N''-Pentamethyldiethylenetriamine 99,0 GC_ASSAY_METHOD

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine

P0881-25ML P0881-500ML P0881-100ML
TCIAP0881-25MLEA 20.9 EUR
TCIAP0881-25ML TCIAP0881-500ML TCIAP0881-100ML
N,N,N',N',N''-Pentamethyldiethylenetriamine 99,0 GC_ASSAY_METHOD
N,N,N',N',N''-Pentamethyldiethylenetriamine
Formule: C₉H₂₃N₃
MW: 173.30 g/mol
Kookpunt: 86°C
Smeltpunt: –20°C
Vlampunt: 72°C
MDL Nummer: MFCD00014876
CAS nummer: 3030-47-5
EINECS: 221-201-1
UN: 2922
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=032d66f35f2a1de03064e377294515ddeb952a24&vid=032d66f35f2a1de03064e377294515ddeb952a24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR