2-(1-Ethylpropyl)pyridine ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P0969-25ML
TCIAP0969-25MLEA 105 EUR
TCIAP0969-25ML
2-(1-Ethylpropyl)pyridine ≥95.0% (by GC)
2-(1-Ethylpropyl)pyridine
Formule: C₁₀H₁₅N
MDL Nummer: MFCD00014631
CAS nummer: 7399-50-0
EINECS: 230-998-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=533aebe523e276c9e31f191dbc131cde69322579&vid=533aebe523e276c9e31f191dbc131cde69322579&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR