Potassium-tert-butoxide ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Potassium tert-butylate, Potassium tert-butanolate

P1008-25G P1008-500G P1008-100G
TCIAP1008-25GEA 19.8 EUR
TCIAP1008-25G TCIAP1008-500G TCIAP1008-100G
Potassium-tert-butoxide ≥97.0% (by titrimetric analysis)
Potassium-tert-butoxide
Formule: C₄H₉KO
Smeltpunt: 256…258 °C
MDL Nummer: MFCD00012162
CAS nummer: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3aa19f1b292300cbf5099f263c66fac165f52ec1&vid=3aa19f1b292300cbf5099f263c66fac165f52ec1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR