Phthalimidoacetaldehyde diethyl acetal ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-(2,2-Diethoxyethyl)isoindoline-1,3-dione, Phthalimidoacetaldehyd Diethylacetal

P1229-25G
TCIAP1229-25GEA 141 EUR
TCIAP1229-25G
Phthalimidoacetaldehyde diethyl acetal ≥95.0% (by GC)
Phthalimidoacetaldehyde diethyl acetal
Formule: C₁₄H₁₇NO₄
MW: 263.30 g/mol
MDL Nummer: MFCD00005901
CAS nummer: 78902-09-7

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 72.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR