2-(Phenylsulphonylmethyl)benzonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

P1456-1G
TCIAP1456-1GEA 94.6 EUR
TCIAP1456-1G
2-(Phenylsulphonylmethyl)benzonitrile ≥98.0% (by GC)
2-(Phenylsulphonylmethyl)benzonitrile
Formule: C₁₄H₁₁NO₂S
MDL Nummer: MFCD06797158
CAS nummer: 82651-72-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Acetonitrile almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR