(Polystyrylmethyl)trimethylammonium cyanoborohydride 2,6 - 3,0 mmol/g, cross-linked with 10% DVB (30-50 mesh)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Benzyltrimethylammonium Cyanoborohydride Resin

P1719-25G P1719-5G
TCIAP1719-25GEA 411 EUR
TCIAP1719-25G TCIAP1719-5G
(Polystyrylmethyl)trimethylammonium cyanoborohydride 2,6 - 3,0 mmol/g, cross-linked with 10% DVB (30-50 mesh)
(Polystyrylmethyl)trimethylammonium cyanoborohydride
MDL Nummer: MFCD02688615
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3536b27d84c43ceabdd4bdd483446de49eb9f7b4&vid=816b8adea5d862669f161d4b15a79198af610578&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Loading 2.6 to 3.0 mmol/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR