Polyethylene glycol monododecyl ether n=∼25 for biochemical research

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

P1777-25G P1777-5G
TCIAP1777-25GEA 83.6 EUR
TCIAP1777-25G TCIAP1777-5G
Polyethylene glycol monododecyl ether n=∼25 for biochemical research
Polyethylene glycol monododecyl ether
Formule: HO(CH₂CH₂O)nC₁₂H₂₅
MDL Nummer: MFCD00080891
CAS nummer: 9002-92-0
EINECS: 500-002-6
UN: 3077
ADR: 9,III
Merck Index: 14,07558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b63b93ffeeac370156291a58fe0e8ed919e66839&vid=f8961edecc6d3e4bcf66e1e8c5cb7ff5665ed258&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to lump
Freezing point 35.0 to 40.0 deg-C
Acid Value max. 1.0
The average value of n 23.0 to 27.0
Suitability for protein-analysis to pass test

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR