2-Phenoxyethyl isobutyrate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P1822-25G P1822-500G
TCIAP1822-25GEA 17.6 EUR
TCIAP1822-25G TCIAP1822-500G
2-Phenoxyethyl isobutyrate ≥98.0% (by GC)
2-Phenoxyethyl isobutyrate
Formule: C₁₂H₁₆O₃
Kookpunt: 127°C
Vlampunt: 113°C
MDL Nummer: MFCD00027363
CAS nummer: 103-60-6
EINECS: 203-127-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=478fb0ee33ce3ee28b21e84f5eaadb1fa88d1693&vid=478fb0ee33ce3ee28b21e84f5eaadb1fa88d1693&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR