2-Pyridyl trifluoromethanesulphonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

P1840-5G
TCIAP1840-5GEA 140 EUR
TCIAP1840-5G
2-Pyridyl trifluoromethanesulphonate ≥98.0% (by GC)
2-Pyridyl trifluoromethanesulphonate
Formule: C₆H₄F₃NO₃S
Kookpunt: 108°C
Vlampunt: 27°C
MDL Nummer: MFCD00192531
CAS nummer: 65007-00-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR