4-(1-Pyrrolidinyl)piperidine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

P1932-25G P1932-5G
TCIAP1932-25GEA 350 EUR
TCIAP1932-25G TCIAP1932-5G
4-(1-Pyrrolidinyl)piperidine ≥98.0% (by GC, titration analysis)
4-(1-Pyrrolidinyl)piperidine
Formule: C₉H₁₈N₂
MDL Nummer: MFCD00038011
CAS nummer: 5004-07-9
EINECS: 225-676-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR