Pazufloxacin mesylate ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P1962-1G P1962-5G
TCIAP1962-1GEA 41.8 EUR
TCIAP1962-1G TCIAP1962-5G
Pazufloxacin mesylate ≥98.0% (by HPLC, titration analysis)
Pazufloxacin mesylate
Formule: C₁₆H₁₅FN₂O₄
Smeltpunt: 259°C
MDL Nummer: MFCD00913262
CAS nummer: 163680-77-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -61.0 to -64.0 deg (C=1, 1mol/L NaOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR