Pyrimidine-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P1993-200MG
TCIAP1993-200MGEA 110 EUR
TCIAP1993-200MG
Pyrimidine-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Pyrimidine-4-carboxylic acid
Formule: C5H4N2O2
MDL Nummer: MFCD00129737
CAS nummer: 31462-59-6
EINECS: 250-641-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cc3963ad969a7c1ca56467f19a05bd075d7e492&vid=2cc3963ad969a7c1ca56467f19a05bd075d7e492&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 225.0 to 229.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR