Penfluridol ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

P2076-200MG P2076-1G
TCIAP2076-200MGEA 72.6 EUR
TCIAP2076-200MG TCIAP2076-1G
Penfluridol ≥98.0% (by titrimetric analysis)
Penfluridol
Formule: C₂₈H₂₇ClF₅NO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00866714
CAS nummer: 26864-56-2
EINECS: 248-074-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,07087
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=865ade52f020c0bf440c7c25de78651f1ba8d8be&vid=865ade52f020c0bf440c7c25de78651f1ba8d8be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR