1-(Phenylsulphonyl)pyrrole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

P2095-1G P2095-5G
TCIAP2095-1GEA 23.1 EUR
TCIAP2095-1G TCIAP2095-5G
1-(Phenylsulphonyl)pyrrole ≥98.0% (by GC)
1-(Phenylsulphonyl)pyrrole
Formule: C₁₀H₉NO₂S
MDL Nummer: MFCD00067739
CAS nummer: 16851-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd78a8bcd791b8cf748c5ad57fbad4a75a1fce70&vid=dd78a8bcd791b8cf748c5ad57fbad4a75a1fce70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR