2-(1H-1,2,4-Triazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P2100-200MG P2100-1G
TCIAP2100-200MGEA 30.8 EUR
TCIAP2100-200MG TCIAP2100-1G
2-(1H-1,2,4-Triazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)
2-(1H-1,2,4-Triazol-3-yl)pyridine
Formule: C₇H₆N₄
MDL Nummer: MFCD13188589
CAS nummer: 23195-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8caa471414525831f602add90fb0733dd55ebd7&vid=c8caa471414525831f602add90fb0733dd55ebd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR