Phenethyl salicylate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Salicylic Acid Phenethyl Ester, 2-Hydroxybenzoic Acid Phenethyl Ester, 2-Hydroxybenzoic Acid 2-Phenylethyl Ester, Salicylic Acid 2-Phenylethyl Ester, Phenethyl 2-Hydroxybenzoate, 2-Phenylethyl 2-Hydroxybenzoate, 2-Phenylethyl Salicylate

P2165-25G
TCIAP2165-25GEA 42.9 EUR
TCIAP2165-25G
Phenethyl salicylate ≥98.0% (by GC)
Phenethyl salicylate
Formule: C₁₅H₁₄O₃
MW: 242.27 g/mol
Kookpunt: 370°C
MDL Nummer: MFCD00020036
CAS nummer: 87-22-9
EINECS: 201-732-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14a027321357c59a7438d30e32181970f8e1bee9&vid=14a027321357c59a7438d30e32181970f8e1bee9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR