Pyridine-2-carboxamidoxime ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

P2590-25G P2590-5G
TCIAP2590-25GEA 125 EUR
TCIAP2590-25G TCIAP2590-5G
Pyridine-2-carboxamidoxime ≥98.0% (by HPLC, titration analysis)
Pyridine-2-carboxamidoxime
Formule: C₆H₇N₃O
MDL Nummer: MFCD00085159
CAS nummer: 1772-01-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR