Pentafluorophenyl Diphenylphosphinate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P2726-1G P2726-5G
TCIAP2726-1GEA 38.5 EUR
TCIAP2726-1G TCIAP2726-5G
Pentafluorophenyl Diphenylphosphinate ≥98.0% (by GC)
Pentafluorophenyl Diphenylphosphinate
Formule: C₁₈H₁₀F₅O₂P
Kookpunt: 153°C
MDL Nummer: MFCD00192380
CAS nummer: 138687-69-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR