Quinaldine red ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 2-(2-(4-Dimethylamino)styryl)-1-ethylquinolinium iodide, 2-(4-(Dimethylamino)styryl)-1-ethylquinolin-1-ium iodide

Q0007-1G Q0007-25G
TCIAQ0007-1GEA 17.6 EUR
TCIAQ0007-1G TCIAQ0007-25G
Quinaldine red ≥95.0% (by titrimetric analysis)
Quinaldine red
Formule: C₂₁H₂₃IN₂
MW: 430.33 g/mol
Smeltpunt: 240
MDL Nummer: MFCD00011968
CAS nummer: 117-92-0
EINECS: 204-221-5
Merck Index: 14,08049
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a759494a820b6172f4c5ca4d511a7f02a77b6608&vid=a759494a820b6172f4c5ca4d511a7f02a77b6608&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Dark green to Dark blue to Black powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Absorbance min. 0.52 (5mg/L methanol, 524 to 528nm)
Transition interval (pH) (colorless to very pale red)1.4 to 3.2 (yellowish red)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR