6-Quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Q0077-25G Q0077-5G
TCIAQ0077-25GEA 308 EUR
TCIAQ0077-25G TCIAQ0077-5G
6-Quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Quinolinecarboxylic acid
Formule: C₁₀H₇NO₂
Kookpunt: 345 °C (1013 hPa)
Smeltpunt: 291°C
MDL Nummer: MFCD00047613
CAS nummer: 10349-57-2
EINECS: 233-761-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b34f2fb29c051b9ad146d062462302a54ecff15&vid=40f3664ea2c6bc68896e6889e7690e3b04ad2dee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR