2-Quinolinecarbonitrile ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Q0099-1G Q0099-5G
TCIAQ0099-1GEA 51.7 EUR
TCIAQ0099-1G TCIAQ0099-5G
2-Quinolinecarbonitrile ≥98.0% (by GC, titration analysis)
2-Quinolinecarbonitrile
Formule: C₁₀H₆N₂
Kookpunt: 160°C
MDL Nummer: MFCD00134341
CAS nummer: 1436-43-7
EINECS: 215-865-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=447b3a66598b2000ddf2739b9cd73fc9e6a48626&vid=447b3a66598b2000ddf2739b9cd73fc9e6a48626&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR