L(+)-Rhamnose monohydrate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal hydrate, 6-Deoxy-L(+)-mannose monohydrate

R0013-25G R0013-5G
TCIAR0013-25GEA 61.6 EUR
TCIAR0013-25G TCIAR0013-5G
L(+)-Rhamnose monohydrate ≥98.0% (by HPLC)
L(+)-Rhamnose monohydrate
Formule: C₆H₁₂O₅·1H₂O
MW: 182.17 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149363
CAS nummer: 10030-85-0
EINECS: 222-793-4
Merck Index: 14,08172
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98ba69487977aa7c90c16eab38619755bae883c1&vid=98ba69487977aa7c90c16eab38619755bae883c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 90.0 to 95.0 deg-C
Specific rotation [a]20/D +8.0 to +8.5 deg (C=10, H2O)[included NH3 aq. (2+3)]
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR