1-(1-Adamantyl)ethylamine hydrochloride ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

R0070-5G
TCIAR0070-5GEA 73.7 EUR
TCIAR0070-5G
1-(1-Adamantyl)ethylamine hydrochloride ≥98.0% (by total nitrogen basis)
1-(1-Adamantyl)ethylamine hydrochloride
Formule: C₁₂H₂₁N
Smeltpunt: 375°C
MDL Nummer: MFCD00072023
CAS nummer: 1501-84-4
Merck Index: 14,08224

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Water almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR