Semicarbazide hydrochloride ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

S0032-25G S0032-500G
TCIAS0032-25GEA 17.6 EUR
TCIAS0032-25G TCIAS0032-500G
Semicarbazide hydrochloride ≥99.0% (by titrimetric analysis)
Semicarbazide hydrochloride
Formule: CH₆ClN₃O
Smeltpunt: 174…178 °C
MDL Nummer: MFCD00013009
CAS nummer: 563-41-7
EINECS: 209-247-0
UN: 2811
ADR: 6.1,III
Merck Index: 14,08444
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d782b35d8db77b8d65f90581338eaa81537c1c32&vid=d782b35d8db77b8d65f90581338eaa81537c1c32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Bromination Method) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR