Succinimide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

S0108-25G S0108-500G
TCIAS0108-25GEA 17.6 EUR
TCIAS0108-25G TCIAS0108-500G
Succinimide ≥98.0% (by titrimetric analysis)
Succinimide
Formule: C₄H₅NO₂
Kookpunt: 130°C
Smeltpunt: 121…124 °C
Dichtheid: 1,418 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005495
CAS nummer: 123-56-8
EINECS: 204-635-6
Merck Index: 14,08871
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=249b7c0490d17949c68ffd63d33817be5b4174ad&vid=249b7c0490d17949c68ffd63d33817be5b4174ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 124.0 to 127.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR