Sudan black B

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB

S0113-25G
TCIAS0113-25GEA 44 EUR
TCIAS0113-25G
Sudan black B
Sudan black B
Formule: C₂₉H₂₄N₆
Smeltpunt: 124°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006919
CAS nummer: 4197-25-5
EINECS: 224-087-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37f8ec1346bc8f7919107d7e9dc6a491109d1e87&vid=37f8ec1346bc8f7919107d7e9dc6a491109d1e87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Dark green to Dark red to Black powder to crystal
Lambda max. 590.0 to 610.0 nm (EtOH)
Absorbance (E1%1cm) min. 630 (EtOH, 590.0 to 610.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR