Sulphanilamide ≥99.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

S0119-25G S0119-500G S0119-100G
TCIAS0119-25GEA 17.6 EUR
TCIAS0119-25G TCIAS0119-500G TCIAS0119-100G
Sulphanilamide ≥99.0% (by HPLC, titration analysis)
Sulphanilamide
Formule: H₂NC₆H₄SO₂NH₂
Kookpunt: 401 °C (1013 hPa)
Smeltpunt: 163…167 °C
Dichtheid: 1,46 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007939
CAS nummer: 63-74-1
EINECS: 200-563-4
Merck Index: 14,08925
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df665336ec0e4fbfe2c0604076fb8298220424e3&vid=df665336ec0e4fbfe2c0604076fb8298220424e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Diazo Method) min. 99.0 %
Solubility in Sodium hydroxide solution almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR