Dibenzyl sebacate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Decanedioic Acid Dibenzyl Ester, Sebacic Acid Dibenzyl Ester

S0147-25G S0147-5G
TCIAS0147-25GEA 135 EUR
TCIAS0147-25G TCIAS0147-5G
Dibenzyl sebacate ≥98.0% (by GC)
Dibenzyl sebacate
Formule: C₂₄H₃₀O₄
MW: 382.50 g/mol
MDL Nummer: MFCD00039640
CAS nummer: 140-24-9
EINECS: 205-404-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d52a8db11c09e0f1048efe2cf83abe451a74300d&vid=d52a8db11c09e0f1048efe2cf83abe451a74300d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR