Lithium stearate ≥90.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

S0237-25G S0237-500G
TCIAS0237-25GEA 26.4 EUR
TCIAS0237-25G TCIAS0237-500G
Lithium stearate ≥90.0% (by titrimetric analysis)
Lithium stearate
Formule: C₁₈H₃₅LiO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042032
CAS nummer: 4485-12-5
EINECS: 224-772-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d3b455d4eaa3a26caa0869a8d46d8e96a68cafb&vid=0d3b455d4eaa3a26caa0869a8d46d8e96a68cafb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystaline
Purity (Nonaqueous Titration) min. 90.0 %
Stearic acid (composition of fatty acid) min. 60.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR