Vinyl stearate ≥95.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

S0241-25G S0241-100G
TCIAS0241-25GEA 24.2 EUR
TCIAS0241-25G TCIAS0241-100G
Vinyl stearate ≥95.0% (by GC) stabilised
Vinyl stearate
Formule: C₂₀H₃₈O₂
MDL Nummer: MFCD00026667
CAS nummer: 111-63-7
EINECS: 203-890-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfe753ea8540473c3f6679c58bba8a95167273e0&vid=bfe753ea8540473c3f6679c58bba8a95167273e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR