Sodium hexafluorophosphate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

S0392-25G
TCIAS0392-25GEA 128 EUR
TCIAS0392-25G
Sodium hexafluorophosphate ≥98.0% (by titrimetric analysis)
Sodium hexafluorophosphate
Formule: F₆NaP
Smeltpunt: 450 °C
Dichtheid: 2,505 g/cm³ (20 °C)
MDL Nummer: MFCD00011122
CAS nummer: 21324-39-0
EINECS: 244-333-1
UN: 3260
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b04d09b4087b98ecac9337c4869a42087d74d9c&vid=1b04d09b4087b98ecac9337c4869a42087d74d9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) 98.0 to 103.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR