Sodium borohydride ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

S0480-100G S0480-500G S0480-25G
TCIAS0480-100GEA 49.5 EUR
TCIAS0480-100G TCIAS0480-500G TCIAS0480-25G
Sodium borohydride ≥95.0% (by titrimetric analysis)
Sodium borohydride
Formule: NaBH₄
Smeltpunt: 400 °C
Dichtheid: 1,07 g/cm³ (20 °C)
Vlampunt: 69 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003518
CAS nummer: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3,I
Merck Index: 14,08592
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53f98a43cd5cd3b0e549ff43068403b1a1ce4ae0&vid=53f98a43cd5cd3b0e549ff43068403b1a1ce4ae0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Sodium hypochlorite Method) min. 95.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR