Succinic acid dihydrazide ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

S0482.25 S0482-500G S0482-100G
TCIAS0482.25EA 37.9 EUR
TCIAS0482.25 TCIAS0482-500G TCIAS0482-100G
Succinic acid dihydrazide ≥97.0% (by HPLC, titration analysis)
Succinic acid dihydrazide
Formule: C₄H₁₀N₄O₂
Smeltpunt: 168°C
MDL Nummer: MFCD00007613
CAS nummer: 4146-43-4
EINECS: 223-970-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62582d5628be6228200972ef63e3624dfe15e3ec&vid=62582d5628be6228200972ef63e3624dfe15e3ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR