Natriumazide ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Sodium triazide

S0489-100G
TCIAS0489-100GEA 36.3 EUR
TCIAS0489-100G
Natriumazide ≥99.0% (by titrimetric analysis)
Natriumazide
Formule: NaN₃
MW: 65.01 g/mol
Smeltpunt: 275
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003536
CAS nummer: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1 II
Merck Index: 14,08581
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=477359b6e50e95e31a2fec931a0b7018253cfc41&vid=477359b6e50e95e31a2fec931a0b7018253cfc41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Cerium redox Method) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR