5-Sulphosalicylic acid dihydrate ≥98.0% (by HPLC, titration analysis) for biochemical research

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

S0830-5G S0830-25G
TCIAS0830-5GEA 17.6 EUR
TCIAS0830-5G TCIAS0830-25G
5-Sulphosalicylic acid dihydrate ≥98.0% (by HPLC, titration analysis) for biochemical research
5-Sulphosalicylic acid dihydrate
Formule: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
Smeltpunt: 113°C
Dichtheid: 1,695 g/cm³ (20 °C)
Vlampunt: 150 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149540
CAS nummer: 5965-83-3
EINECS: 202-555-6
ADR: 8,III
Merck Index: 14,08964
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bbb92f5f3dd496ec3e92811bf21e76905eb29d8&vid=16960336942131c0215190e82428597dff086160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Protein precipitation test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR