D-(-)-Tartaric acid ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T0026-500G T0026-25G T0026-100G
TCIAT0026-500GEA 270 EUR
TCIAT0026-500G TCIAT0026-25G TCIAT0026-100G
D-(-)-Tartaric acid ≥99.0% (by titrimetric analysis)
D-(-)-Tartaric acid
Formule: C₄H₆O₆
Kookpunt: 356 °C (1013 hPa)
Smeltpunt: 166…169 °C
Dichtheid: 1,76 g/cm³ (20 °C)
Vlampunt: 210 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004238
CAS nummer: 147-71-7
EINECS: 205-695-6
Merck Index: 14,09068
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fddb94106dbe15d622d983c89ed728767d3b8c5d&vid=fddb94106dbe15d622d983c89ed728767d3b8c5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Specific rotation [a]20/D -11.5 to -13.0 deg (C=20, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR