Tetrabromobisphenol A ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T0032-25G T0032-500G
TCIAT0032-25GEA 18.7 EUR
TCIAT0032-25G TCIAT0032-500G
Tetrabromobisphenol A ≥98.0% (by GC, titration analysis)
Tetrabromobisphenol A
Formule: C₁₅H₁₂Br₄O₂
MDL Nummer: MFCD00013962
CAS nummer: 79-94-7
EINECS: 201-236-9
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4528bba6abf6997e4744b344b339dbc4686ce32&vid=a4528bba6abf6997e4744b344b339dbc4686ce32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 178.0 to 184.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR