Eosin Y 95,0 HPLC, T

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-

T0035-25G
TCIAT0035-25GEA 51.7 EUR
TCIAT0035-25G
Eosin Y 95,0 HPLC, T
Eosin Y
Formule: C₂₀H₈Br₄O₅
MW: 647.89 g/mol
Smeltpunt: 295
MDL Nummer: MFCD00036189
CAS nummer: 15086-94-9
EINECS: 239-138-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27cf603f6b053d3ffdda06a2489522fbc9dfd9bb&vid=27cf603f6b053d3ffdda06a2489522fbc9dfd9bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange to Red powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 93.0 %
Lambda max. 515.0 to 519.0 nm (0.1mol/L NaOH sol.)
Absorbance (E1%1cm) min. 1,000 (0.1mol/L NaOH sol., 515.0 to 519.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR