Tetramethylammonium hydroxide 10% in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T0138-25ML T0138-500ML T0138-100ML
TCIAT0138-25MLEA 19.8 EUR
TCIAT0138-25ML TCIAT0138-500ML TCIAT0138-100ML
Tetramethylammonium hydroxide 10% in water
Tetramethylammonium hydroxide
Formule: C₄H₁₃NO
Kookpunt: 110 °C (1013 hPa)
Dichtheid: 1,02 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008280
CAS nummer: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8,II
Merck Index: 14,09224
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c995cdbad5279cf8f8fe6dba9fac2429eed1f9d9&vid=e8a578d1a23b1c22d6eca40cbe634af315f64dfa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (Neutralization titration) 10.0 to 11.0 w/w%
Ignition residue (Sulfate) max. 0.1 %
Chloride max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR