Tetramethylthiurammonosulfide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T0157-25G T0157-500G
TCIAT0157-25GEA 17.6 EUR
TCIAT0157-25G TCIAT0157-500G
Tetramethylthiurammonosulfide ≥98.0% (by GC)
Tetramethylthiurammonosulfide
Formule: C₆H₁₂N₂S₃
Smeltpunt: 110 °C
Dichtheid: 1,37 g/cm³ (20 °C)
MDL Nummer: MFCD00014870
CAS nummer: 97-74-5
EINECS: 202-605-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed63a98b8d44c380a166c098b952d4180896a1df&vid=ed63a98b8d44c380a166c098b952d4180896a1df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystaline
Purity (GC) min. 98.0 %
Melting point 108.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR