Tetrapropylammonium hydroxide 10% in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T0171-25ML T0171-100ML
TCIAT0171-25MLEA 30.8 EUR
TCIAT0171-25ML TCIAT0171-100ML
Tetrapropylammonium hydroxide 10% in water
Tetrapropylammonium hydroxide
Formule: C₁₂H₂₉NO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009360
CAS nummer: 4499-86-9
EINECS: 224-800-6
UN: 3267
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b00842875850afe13ae3d2a04e88d93832049157&vid=3de79e0cc124a3efb5a87da77a818989f7df6cc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (Neutralization titration) 10.0 to 11.0 w/w%
Bromide max. 0.10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR