Cocarboxylase ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T0183-5G
TCIAT0183-5GEA 136 EUR
TCIAT0183-5G
Cocarboxylase ≥98.0% (by titrimetric analysis)
Cocarboxylase
Formule: C₁₂H₁₉ClN₄O₇P₂S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00038740
CAS nummer: 154-87-0
EINECS: 205-836-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eba29fbd9bfa4533a336a146937f9246f512af2&vid=6eba29fbd9bfa4533a336a146937f9246f512af2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR