2-Thiobarbituric acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T0192-500G T0192-25G T0192-100G
TCIAT0192-500GEA 607 EUR
TCIAT0192-500G TCIAT0192-25G TCIAT0192-100G
2-Thiobarbituric acid ≥98.0% (by titrimetric analysis)
2-Thiobarbituric acid
Formule: C₄H₄N₂O₂S
Kookpunt: 419 °C (1013 hPa)
Smeltpunt: 245°C
Dichtheid: 1,661 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006674
CAS nummer: 504-17-6
EINECS: 207-985-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2aa4c59b5256ea1169b20f1dcec5715f4f7e2ba&vid=a2aa4c59b5256ea1169b20f1dcec5715f4f7e2ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Sodium hydroxide solution almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR