4,4'-Bi-o-toluidine dihydrochloride ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T0254-25G
TCIAT0254-25GEA 48.4 EUR
TCIAT0254-25G
4,4'-Bi-o-toluidine dihydrochloride ≥98.0% (by titrimetric analysis)
4,4'-Bi-o-toluidine dihydrochloride
Formule: C₁₄H₁₈Cl₂N₂
Smeltpunt: 130 °C
MDL Nummer: MFCD00012960
CAS nummer: 612-82-8
EINECS: 210-322-5
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b589a8bca6bd046e52e349a48d878780df635619&vid=b589a8bca6bd046e52e349a48d878780df635619&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Red powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency
Sensitiveness to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR