Tolueen ≥99.5% (by GC) for spectrophotometry

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T0260-100ML
TCIAT0260-100MLEA 28.6 EUR
TCIAT0260-100ML
Tolueen ≥99.5% (by GC) for spectrophotometry
Tolueen
Formule: C₆H₅CH₃
Kookpunt: 110°C (literatuur)
Dichtheid: 0,867 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
Bereik: 05-2114615130-69
Merck Index: 14,09529
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b212b27aba5aa115b97760350023c6228bd4cf96&vid=5f88e5bd763706d85b70fe26a6cc2d89ab0d7670&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 99.5 %
Absorbance (290nm) max. 0.350
Absorbance (300nm) max. 0.150
Absorbance (310nm) max. 0.080
Absorbance(300nm) max. 0.150
Absorbance(310nm) max. 0.080
Absorbance(320nm) max. 0.050
Absorbance(330nm) max. 0.020
Absorbance(340nm) max. 0.010
Absorbance(400nm) max. 0.010
Water max. 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR