Tricosanoic acid ≥95.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T0402-1G T0402-10G
TCIAT0402-1GEA 82.5 EUR
TCIAT0402-1G TCIAT0402-10G
Tricosanoic acid ≥95.0% (by GC, titration analysis)
Tricosanoic acid
Formule: C₂₃H₄₆O₂
MW: 354.62 g/mol
Kookpunt: 399
Smeltpunt: 76
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002808
CAS nummer: 2433-96-7
EINECS: 219-419-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e014916fbf1b5af803956db0e084bd95a444815&vid=8fb641c38c990f35e8fa180ff0a1734870606891&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 78.0 to 81.0 deg-C
Solubility in hot EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR